Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012522.2 | NP_036654.2 | 3 | 424 | ||
GenBank mRNA | D01098.1 | - | 3 | 425 | ||
FQ211441.1 | - | 3 | 483 | |||
M88344.1 | - | 3 | 573 | |||
M88346.1 | - | 2 | 319 | |||
M88347.1 | - | 2 | 319 |