Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cela1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012552.3 | NP_036684.1 | 4, 5 | 357 | ||
GenBank mRNA | FQ227752.1 | - | 4, 5 | 379 | ||
V01234.1 | - | 4, 5 | 378 |