Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012580.2 | NP_036712.1 | 3 | 313 | ||
GenBank mRNA | BC091164.1 | - | 3 | 313 | 7384448 | |
FQ228696.1 | - | 2 | 187 |