Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012581.2 | NP_036713.2 | 2 | 768 | ||
GenBank mRNA | M91802.1 | - | 2 | 805 |
Gene Symbol: NEWGENE_2813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762936.1 | XP_008761158.1 | 2 | 1349 |