Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177305.1 | NP_001170776.1 | 9 | 1091 | ||
NM_001271536.1 | NP_001258465.1 | 9 | 1177 | |||
NM_012495.2 | NP_036627.1 | 9 | 1193 | |||
XM_006230209.2 | XP_006230271.1 | 11 | 1439 | |||
XM_006230210.2 | XP_006230272.1 | 10 | 1349 | |||
XM_006230211.1 | XP_006230273.1 | 10 | 1204 | |||
GenBank mRNA | BC064440.1 | - | 9 | 1170 | 5623628 | |
CO383722.1 | - | 2 | 198 | |||
FQ212782.1 | - | 9 | 1144 | |||
FQ213869.1 | - | 9 | 1176 | |||
FQ214123.1 | - | 9 | 1144 | |||
FQ214510.1 | - | 9 | 1146 | |||
FQ214648.1 | - | 5 | 565 | |||
FQ215258.1 | - | 5 | 565 | |||
FQ215311.1 | - | 5 | 565 | |||
FQ215443.1 | - | 5 | 564 | |||
FQ215968.1 | - | 5 | 565 | |||
FQ216189.1 | - | 9 | 1099 | |||
FQ216413.1 | - | 5 | 566 | |||
FQ216588.1 | - | 5 | 566 | |||
FQ217110.1 | - | 5 | 565 | |||
FQ217473.1 | - | 5 | 565 | |||
FQ217887.1 | - | 5 | 560 | |||
FQ223546.1 | - | 3 | 261 | |||
FQ224221.1 | - | 9 | 1185 | |||
FQ224440.1 | - | 2 | 235 | |||
FQ224579.1 | - | 2 | 188 | |||
FQ228954.1 | - | 9 | 1123 | |||
FQ229100.1 | - | 9 | 1141 | |||
M12919.1 | - | 9 | 1193 | |||
M14420.1 | - | 9 | 1091 | |||
M28282.1 | - | 2 | 60 |