Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012544.1 | NP_036676.1 | 19 | 2805 | ||
GenBank mRNA | AF201331.1 | - | 19 | 2805 | ||
AF201332.1 | - | 19 | 2805 | |||
AF532783.1 | - | 19 | 2749 | |||
AF532784.1 | - | 19 | 2749 | |||
AF539425.1 | - | 1182 | ||||
BC085760.1 | - | 19 | 2805 | 7097136 | ||
U03708.1 | - | 19 | 2805 | |||
U03734.1 | - | 19 | 2805 |