Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012548.2 | NP_036680.1 | 4 | 612 | ||
GenBank mRNA | M64711.1 | - | 4 | 607 |