Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012557.2 | NP_036689.2 | 8 | 1070 | ||
XM_006253488.2 | XP_006253550.1 | 9 | 1345 | |||
XM_006253489.2 | XP_006253551.1 | 8 | 1202 | |||
XM_006253490.2 | XP_006253552.1 | 8 | 1047 | |||
XM_006253491.2 | XP_006253553.1 | 5 | 1876 | |||
XM_008771423.1 | XP_008769645.1 | 7 | 2192 | |||
GenBank mRNA | BC168147.1 | - | 8 | 1070 | 9024689 | |
U73586.1 | - | 7 | 846 |