Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012564.3 | NP_036696.2 | 10 | 1238 | ||
GenBank mRNA | BC078891.1 | - | 10 | 1238 | 7130137 | |
FQ209578.1 | - | 10 | 1187 | |||
FQ209652.1 | - | 10 | 1249 | |||
FQ209886.1 | - | 10 | 1248 | |||
FQ210204.1 | - | 10 | 952 | |||
FQ210565.1 | - | 3 | 332 | |||
FQ210734.1 | - | 10 | 1246 | |||
FQ219041.1 | - | 7 | 859 | |||
FQ219081.1 | - | 10 | 1249 | |||
FQ219132.1 | - | 6 | 679 | |||
FQ219502.1 | - | 10 | 1246 | |||
J05148.1 | - | 10 | 1214 | |||
M12450.1 | - | 10 | 1194 |