Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320979.1 | NP_001307908.1 | 6 | 1095 | ||
NM_003689.3 | NP_003680.2 | 7 | 1200 | |||
GenBank mRNA | AF026947.1 | - | 7 | 1168 | ||
BC004111.2 | - | 7 | 1127 | 3832140 | ||
BC007352.2 | - | 7 | 1175 | |||
BC010852.1 | - | 7 | 1176 | 3898226 | ||
BC011586.1 | - | 7 | 1127 | 4620484 | ||
BC012171.1 | - | 7 | 1175 | 4642811 | ||
BC013996.2 | - | 7 | 1131 | 3625337 | ||
BU944847.1 | - | 1 | 271 | |||
EU794591.1 | - | 7 | 1200 | |||
Y16675.1 | - | 7 | 1162 |