Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012613.1 | NP_036745.1 | 1 | 813 | ||
XM_006232591.2 | XP_006232653.1 | 2 | 1314 | |||
GenBank mRNA | BC128742.1 | - | 1 | 799 | 7458324 | |
M74535.1 | - | 2 | 762 | |||
X14773.1 | - | 1 | 670 |