Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012647.1 | NP_036779.1 | 2 | 344 | ||
GenBank mRNA | M22254.1 | - | 2 | 344 | ||
X03639.1 | - | 2 | 344 |