Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000291.3 | NP_000282.1 | 3 | 303 | ||
GenBank mRNA | AB062432.1 | - | 3 | 200 | ||
AK291081.1 | - | 3 | 217 | |||
AK295014.1 | - | 3 | 217 | |||
AK295182.1 | - | 3 | 217 | |||
AK298855.1 | - | 3 | 217 | |||
AK301632.1 | - | 3 | 226 | |||
AK301740.1 | - | 3 | 376 | |||
AK310756.1 | - | 3 | 878 | |||
AK312280.1 | - | 3 | 217 | |||
AY423725.1 | - | 3 | 153 | |||
BC023234.1 | - | 3 | 167 | 3917985 | ||
BC103752.1 | - | 3 | 217 | 6526214 | ||
BC104837.1 | - | 3 | 182 | 8143840 | ||
BC113568.1 | - | 3 | 182 | 8322620 | ||
CR456716.1 | - | 3 | 131 | |||
DC336483.1 | - | 3 | 303 | |||
EU794645.1 | - | 3 | 217 | |||
L00160.1 | - | 2 | 66 | |||
V00572.1 | - | 3 | 210 |