Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012611.3 | NP_036743.3 | 321 | |||
GenBank mRNA | AY211532.1 | - | 132 | |||
D12520.1 | - | 321 | ||||
D14051.1 | - | 151 | ||||
D44591.1 | - | 152 | ||||
D83661.1 | - | 131 | ||||
L12562.1 | - | 233 | ||||
U03699.1 | - | 134 | ||||
U26686.1 | - | 131 | ||||
X76881.1 | - | 316 |
Gene Symbol: LOC497963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246949.2 | XP_006247011.1 | 186 |