Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100912228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749745.3 | XP_003749793.1 | 386 |
Gene Symbol: Npy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012614.2 | NP_036746.1 | 3, 4 | 373 | ||
GenBank mRNA | JX157883.1 | - | 3, 4 | 194 | ||
M15880.1 | - | 3, 4 | 348 | |||
M20373.1 | - | 3, 4 | 328 |