Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012664.2 | NP_036796.1 | 2, 3 | 405 | ||
GenBank mRNA | BC099798.1 | - | 420 | 7936715 | ||
X06177.1 | - | 414 | ||||
X06388.1 | - | 446 | ||||
X06655.1 | - | 378 |