Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012676.1 | NP_036808.1 | 4, 5 | 176 | ||
XM_006249822.1 | XP_006249884.1 | 6, 7 | 285 | |||
XM_006249823.1 | XP_006249885.1 | 6, 7 | 286 | |||
XM_006249825.2 | XP_006249887.1 | 6, 7 | 299 | |||
XM_006249828.1 | XP_006249890.1 | 5, 6 | 273 | |||
XM_006249829.1 | XP_006249891.1 | 5, 6 | 274 | |||
XM_006249833.2 | XP_006249895.1 | 5, 6 | 281 | |||
XM_006249835.1 | XP_006249897.1 | 4, 5 | 243 | |||
XM_006249836.1 | XP_006249898.1 | 4, 5 | 244 | |||
XM_006249838.1 | XP_006249900.1 | 4, 5 | 244 | |||
XM_006249840.2 | XP_006249902.1 | 7, 8 | 315 | |||
XM_006249842.2 | XP_006249904.1 | 5, 6 | 272 | |||
XM_008769535.1 | XP_008767757.1 | 6, 7 | 286 | |||
XM_008769536.1 | XP_008767758.1 | 6, 7 | 286 | |||
XM_008769537.1 | XP_008767759.1 | 5, 6 | 274 | |||
XM_008769538.1 | XP_008767760.1 | 4, 5 | 244 | |||
GenBank mRNA | BC161855.1 | - | 5, 6 | 281 | 9035753 | |
EU295527.1 | - | 3, 4 | 146 | |||
M26051.1 | - | 4, 5 | 176 | |||
M26052.1 | - | 4, 5 | 196 |