Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012677.1 | NP_036809.1 | 4 | 647 | ||
GenBank mRNA | M11565.1 | - | 4 | 647 |