Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081955.2 | NP_001075424.1 | 13 | 1136 | ||
NM_001165933.1 | NP_001159405.1 | 13 | 1136 | |||
NM_003835.3 | NP_003826.2 | 13 | 1145 | |||
XM_011525426.2 | XP_011523728.1 | 6 | 1316 | |||
GenBank mRNA | AF071476.1 | - | 13 | 1009 | ||
AF073710.1 | - | 4 | 870 | |||
AF493932.1 | - | 3 | 255 | |||
AF493933.1 | - | 13 | 942 | |||
AK289876.1 | - | 13 | 1009 | |||
AK290535.1 | - | 13 | 1005 | |||
AK301719.1 | - | 13 | 1015 | |||
AK309061.1 | - | 13 | 1026 | |||
AY585190.1 | - | 3 | 255 | |||
AY585191.1 | - | 13 | 933 | |||
BC022504.1 | - | 13 | 1003 | 4801328 | ||
BC096843.1 | - | 3 | 258 | 6519202 |