Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpine1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012620.1 | NP_036752.1 | 3, 4 | 631 | ||
GenBank mRNA | M24067.1 | - | 3, 4 | 631 |