Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012657.2 | NP_036789.2 | 3, 4 | 1007 | ||
XM_006240458.2 | XP_006240520.1 | 3, 4 | 1050 | |||
GenBank mRNA | BC062236.1 | - | 3, 4 | 1011 | 6888893 | |
CO560755.1 | - | 2, 3 | 502 | |||
D00751.1 | - | 3, 4 | 960 | |||
FQ209577.1 | - | 3, 4 | 1009 | |||
FQ218085.1 | - | 3, 4 | 1012 | |||
FQ218647.1 | - | 2, 3 | 684 | |||
FQ218802.1 | - | 3, 4 | 1009 | |||
FQ219296.1 | - | 3, 4 | 1013 | |||
FQ219312.1 | - | 3, 4 | 1011 | |||
M15916.1 | - | 3, 4 | 972 | |||
X05348.1 | - | 3, 4 | 994 | |||
X16358.1 | - | 3, 4 | 1008 |