Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012675.3 | NP_036807.1 | 4 | 833 | ||
GenBank mRNA | AF269159.1 | - | 4 | 689 | ||
AF269160.1 | - | 4 | 689 | |||
AJ002278.1 | - | 4 | 689 | |||
BC107671.1 | - | 4 | 833 | 7938033 | ||
X66539.1 | - | 4 | 689 |
Gene Symbol: LOC103694380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772775.1 | XP_008770997.1 | 4 | 844 |