Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cckar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012688.3 | NP_036820.1 | 5 | 994 | ||
XM_008770188.1 | XP_008768410.1 | 6 | 1178 | |||
GenBank mRNA | M88096.1 | - | 5 | 998 |
Set Membership: |
G-protein coupled receptor |