Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172305.1 | NP_001165776.1 | 1 | 237 | ||
NM_012713.3 | NP_036845.3 | 1 | 237 | |||
GenBank mRNA | M13706.1 | - | 1 | 247 | ||
M19007.1 | - | 1 | 197 | |||
X04440.1 | - | 1 | 791 |