Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012753.2 | NP_036885.1 | 1 | 209 | ||
XM_006231434.2 | XP_006231496.1 | 2 | 532 | |||
XM_006231435.2 | XP_006231497.1 | 2 | 501 | |||
GenBank mRNA | BC078898.1 | - | 1 | 209 | 7111017 | |
M22204.1 | - | 1 | 211 | |||
M31681.1 | - | 1 | 221 | |||
X14086.1 | - | 1 | 211 |