Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012754.1 | NP_036886.1 | 3 | 791 | ||
XM_006240221.2 | XP_006240283.1 | 3 | 1032 | |||
GenBank mRNA | AB012721.1 | - | 2 | 375 | ||
AB190769.1 | - | 2 | 614 | |||
AB190770.1 | - | 2 | 614 | |||
AF042058.1 | - | 2 | 375 | |||
AF042059.1 | - | 2 | 375 | |||
AF042060.1 | - | 2 | 375 | |||
AF042061.1 | - | 2 | 375 | |||
AF161187.1 | - | 1 | 87 | |||
U57439.1 | - | 3 | 791 |