Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012762.2 | NP_036894.2 | 1 | 119 | ||
GenBank mRNA | D85899.1 | - | 1 | 119 | ||
U14647.1 | - | 1 | 119 |