Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003883.3 | NP_003874.2 | 3 | 266 | ||
GenBank mRNA | AF005482.1 | - | 1 | 282 | ||
AF039703.1 | - | 3 | 222 | |||
BC000614.1 | - | 3 | 255 | 3345755 | ||
BM461585.1 | - | 3 | 256 | |||
DB456387.1 | - | 3 | 280 | |||
U66914.1 | - | 3 | 211 | |||
U75696.1 | - | 2 | 249 | |||
U75697.1 | - | 3 | 255 |