Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206897.1 | NP_001193826.1 | 4 | 665 | ||
NM_003888.3 | NP_003879.2 | 3 | 568 | |||
NM_170696.2 | NP_733797.1 | 3 | 568 | |||
NM_170697.2 | NP_733798.1 | 1 | 172 | |||
GenBank mRNA | AB015226.1 | - | 3 | 338 | ||
AB015227.1 | - | 1 | 101 | |||
AB015228.1 | - | 1 | 63 | |||
AK128709.1 | - | 3 | 1063 | |||
AK303057.1 | - | 4 | 410 | |||
BC030589.2 | - | 3 | 326 | 4826743 | ||
BX443600.2 | - | 1 | 172 |