Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012786.3 | NP_036918.2 | 2 | 228 | ||
GenBank mRNA | FM054348.1 | - | 2 | 228 | ||
X64827.1 | - | 2 | 223 |