Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Proc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012803.1 | NP_036935.1 | 3 | 236 | ||
XM_006254526.2 | XP_006254588.1 | 3 | 308 | |||
XM_006254527.2 | XP_006254589.1 | 3 | 315 | |||
XM_008772018.1 | XP_008770240.1 | 3 | 378 | |||
GenBank mRNA | BC078879.1 | - | 2 | 213 | 7122006 | |
X64336.1 | - | 3 | 236 |