Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012736.1 | NP_036868.1 | 3, 4 | 373 | ||
GenBank mRNA | BC083565.1 | - | 3, 4 | 331 | 7134834 | |
FQ223746.1 | - | 629 | ||||
U08027.1 | - | 3, 4 | 352 | |||
X78593.1 | - | 3, 4 | 373 |