Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012759.2 | NP_036891.2 | 2, 3 | 374 | ||
GenBank mRNA | BC091160.1 | - | 2, 3 | 374 | 7373783 | |
U39476.1 | - | 2, 3 | 373 |