Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161441.1 | NP_001154913.1 | 6 | 774 | ||
NM_001161442.1 | NP_001154914.1 | 6 | 713 | |||
NM_001161443.1 | NP_001154915.1 | 5 | 623 | |||
NM_001161444.1 | NP_001154916.1 | 6 | 744 | |||
NM_003975.3 | NP_003966.2 | 6 | 744 | |||
XM_006711615.2 | XP_006711678.1 | 6 | 762 | |||
XM_017002762.1 | XP_016858251.1 | 6 | 791 | |||
XM_017002763.1 | XP_016858252.1 | 6 | 782 | |||
XM_017002764.1 | XP_016858253.1 | 6 | 729 | |||
XM_017002765.1 | XP_016858254.1 | 6 | 742 | |||
XM_017002766.1 | XP_016858255.1 | 6 | 732 | |||
XM_017002767.1 | XP_016858256.1 | 6 | 797 | |||
GenBank mRNA | AF051325.1 | - | 6 | 666 | ||
AF097744.1 | - | 6 | 643 | |||
AJ000553.1 | - | 6 | 690 | |||
AK222737.1 | - | 6 | 667 | |||
AK313706.1 | - | 6 | 738 | |||
AY763098.1 | - | 5 | 623 | |||
AY763099.1 | - | 6 | 720 | |||
AY763100.1 | - | 6 | 650 | |||
BC012107.1 | - | 6 | 706 | 4636736 |