Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012796.2 | NP_036928.1 | 4 | 529 | ||
GenBank mRNA | BC061856.1 | - | 4 | 533 | 5623709 | |
CK473092.1 | - | 4 | 529 | |||
D10026.1 | - | 4 | 517 | |||
FJ179409.1 | - | 4 | 395 |
Gene Symbol: LOC103694879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773848.1 | XP_008772070.1 | 2 | 599 |