Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012707.2 | NP_036839.2 | 4 | 414 | ||
GenBank mRNA | BP479145.1 | - | 4 | 414 | ||
BP480555.1 | - | 4 | 389 |