Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012738.1 | NP_036870.1 | 4 | 780 | ||
GenBank mRNA | BC089820.1 | - | 3 | 764 | 7321674 | |
FQ209434.1 | - | 4 | 781 | |||
FQ209776.1 | - | 5 | 736 | |||
FQ209818.1 | - | 4 | 786 | |||
FQ209900.1 | - | 4 | 781 | |||
FQ209928.1 | - | 4 | 785 | |||
FQ209958.1 | - | 4 | 785 | |||
FQ210010.1 | - | 4 | 785 | |||
FQ210126.1 | - | 4 | 782 | |||
FQ210161.1 | - | 4 | 808 | |||
FQ210244.1 | - | 1 | 442 | |||
FQ210354.1 | - | 4 | 785 | |||
FQ210371.1 | - | 4 | 782 | |||
FQ210470.1 | - | 4 | 781 | |||
FQ210544.1 | - | 4 | 783 | |||
FQ210653.1 | - | 4 | 783 | |||
FQ210722.1 | - | 4 | 785 | |||
FQ210732.1 | - | 1 | 422 | |||
FQ210863.1 | - | 4 | 785 | |||
FQ210880.1 | - | 4 | 781 | |||
FQ210914.1 | - | 4 | 781 | |||
FQ211297.1 | - | 4 | 783 | |||
FQ211535.1 | - | 4 | 788 | |||
FQ211571.1 | - | 4 | 781 | |||
FQ218078.1 | - | 4 | 781 | |||
FQ218079.1 | - | 4 | 785 | |||
FQ218325.1 | - | 4 | 785 | |||
FQ218519.1 | - | 4 | 781 | |||
FQ218663.1 | - | 4 | 781 | |||
FQ218837.1 | - | 4 | 787 | |||
FQ219021.1 | - | 4 | 785 | |||
FQ219074.1 | - | 4 | 781 | |||
FQ219479.1 | - | 4 | 873 | |||
FQ219515.1 | - | 4 | 785 | |||
FQ219577.1 | - | 4 | 781 | |||
FQ219728.1 | - | 4 | 780 | |||
FQ220986.1 | - | 4 | 782 | |||
FQ224655.1 | - | 4 | 781 | |||
FQ229884.1 | - | 4 | 820 | |||
M00001.1 | - | 4 | 780 | |||
X00558.1 | - | 3 | 769 |