Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012757.2 | NP_036889.1 | 4 | 1458 | ||
XM_006227859.2 | XP_006227921.1 | 3 | 2720 | |||
XM_006227860.2 | XP_006227922.1 | 3 | 2842 | |||
XM_006227867.2 | XP_006227929.1 | 2 | 2531 | |||
XM_006227868.2 | XP_006227930.1 | 2 | 2479 | |||
XM_008758726.1 | XP_008756948.1 | 3 | 2839 | |||
XM_008758727.1 | XP_008756949.1 | 3 | 2790 | |||
XM_008758728.1 | XP_008756950.1 | 3 | 2486 | |||
XM_008758729.1 | XP_008756951.1 | 5 | 1775 | |||
XM_008758730.1 | XP_008756952.1 | 3 | 2787 | |||
GenBank mRNA | BC078884.1 | - | 4 | 1458 | 7131760 |