Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012781.3 | NP_036913.3 | 9 | 1011 | ||
XM_006229494.2 | XP_006229556.1 | 10 | 1727 | |||
XM_006229495.2 | XP_006229557.1 | 9 | 1820 | |||
XM_006229496.2 | XP_006229558.1 | 9 | 2281 | |||
XM_006229497.2 | XP_006229559.1 | 9 | 1664 | |||
XM_006229498.2 | XP_006229560.1 | 9 | 962 | |||
XM_008759602.1 | XP_008757824.1 | 9 | 1380 | |||
XM_008759603.1 | XP_008757825.1 | 9 | 1369 | |||
XM_008759604.1 | XP_008757826.1 | 9 | 4100 | |||
XM_008759605.1 | XP_008757827.1 | 9 | 1294 | |||
XM_008759606.1 | XP_008757828.1 | 9 | 1519 | |||
XM_008759607.1 | XP_008757829.1 | 9 | 1076 | |||
XM_008759608.1 | XP_008757830.1 | 8 | 1176 | |||
GenBank mRNA | AY264366.1 | - | 9 | 940 | ||
DQ223732.1 | - | 1011 | ||||
U61157.1 | - | 9 | 1035 | |||
U61405.1 | - | 9 | 1011 |