Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012791.2 | NP_036923.1 | 4, 5 | 462 | ||
XM_006248031.2 | XP_006248093.1 | 4, 5 | 3407 | |||
XM_008768562.1 | XP_008766784.1 | 3, 4 | 2670 | |||
XM_008768563.1 | XP_008766785.1 | 5, 6 | 3543 | |||
XM_008768564.1 | XP_008766786.1 | 4, 5 | 509 | |||
XM_008768565.1 | XP_008766787.1 | 4, 5 | 584 | |||
GenBank mRNA | X79769.1 | - | 4, 5 | 464 |