Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012795.2 | NP_036927.1 | 2 | 1719 | ||
XM_006248483.2 | XP_006248545.1 | 2 | 2005 |
Gene Symbol: LOC100911551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751076.3 | XP_003751124.2 | 1 | 1717 |