Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260486.1 | NP_001247415.1 | 5 | 617 | ||
NM_001260487.1 | NP_001247416.1 | 5 | 500 | |||
NM_001260488.1 | NP_001247417.1 | 6 | 603 | |||
NM_003445.3 | NP_003436.3 | 5 | 591 | |||
NM_198089.2 | NP_932355.2 | 5 | 514 | |||
XM_005259215.3 | XP_005259272.1 | 5 | 661 | |||
XM_011527278.2 | XP_011525580.1 | 6 | 546 | |||
XM_011527279.2 | XP_011525581.1 | 6 | 1606 | |||
XM_017027248.1 | XP_016882737.1 | 6 | 638 | |||
GenBank mRNA | AF187986.1 | - | 5 | 544 | ||
AK022875.1 | - | 5 | 481 | |||
AK297360.1 | - | 6 | 579 | |||
AK297384.1 | - | 4 | 327 | |||
AK314510.1 | - | 5 | 496 | |||
BC075050.2 | - | 4 | 385 | 30915579 | ||
BC075051.2 | - | 4 | 385 | 30915363 | ||
BC126377.1 | - | 4 | 439 | 8992093 | ||
BC130381.1 | - | 4 | 439 | 40146412 | ||
BQ893627.1 | - | 5 | 472 | |||
DA111916.1 | - | 5 | 571 | |||
DC330742.1 | - | 5 | 479 | |||
DN994146.1 | - | 5 | 521 |