Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012832.3 | NP_036964.3 | 2 | 101 | ||
XM_008759498.1 | XP_008757720.1 | 2 | 161 | |||
GenBank mRNA | AY574256.1 | - | 2 | 101 | ||
AY671974.1 | - | 2 | 104 | |||
L31619.1 | - | 2 | 123 | |||
S53987.1 | - | 142 |