Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012836.2 | NP_036968.1 | 1, 2 | 793 | ||
GenBank mRNA | U62897.1 | - | 1, 2 | 793 |