Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012849.1 | NP_036981.1 | 3 | 296 | ||
GenBank mRNA | M38653.1 | - | 3 | 296 |