Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012874.1 | NP_037006.1 | 2, 3 | 574 | ||
XM_008772862.1 | XP_008771084.1 | 2, 3 | 755 | |||
XM_008772863.1 | XP_008771085.1 | 2, 3 | 290 | |||
GenBank mRNA | M35104.1 | - | 2, 3 | 574 | ||
M35105.1 | - | 2, 3 | 574 | |||
M35106.1 | - | 2, 3 | 574 |