Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012920.1 | NP_037052.1 | 2 | 160 | ||
GenBank mRNA | J02942.1 | - | 2 | 160 |