Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012861.1 | NP_036993.1 | 4 | 405 | ||
GenBank mRNA | M76704.1 | - | 3 | 350 | ||
S61804.1 | - | 4 | 405 | |||
X54862.1 | - | 4 | 431 |