Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012914.1 | NP_037046.1 | 5 | 490 | ||
XM_006246597.2 | XP_006246659.1 | 6 | 1104 | |||
GenBank mRNA | M30581.1 | - | 5 | 490 |
Gene Symbol: LOC102549389 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768230.1 | XP_008766452.1 | 2 | 128 |