Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008895.2 | NP_001008895.1 | 7 | 877 | ||
NM_001278513.1 | NP_001265442.1 | 7 | 1021 | |||
NM_001278514.1 | NP_001265443.1 | 7 | 689 | |||
NM_003589.2 | NP_003580.1 | 7 | 703 | |||
XM_011537523.2 | XP_011535825.1 | 6 | 1237 | |||
GenBank mRNA | AB012193.1 | - | 1 | 51 | ||
AB178950.1 | - | 7 | 754 | |||
AF077188.1 | - | 7 | 614 | |||
AK126278.1 | - | 2 | 1086 | |||
AK314835.1 | - | 7 | 976 | |||
AL833355.1 | - | 6 | 641 | |||
AY365124.1 | - | 7 | 754 | |||
BC008308.1 | - | 7 | 765 | 3537176 |